3-Amino-4-hydroxybenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-Amino-4-hydroxybenzonitrile, 2-Amino-4-cyanophenol, 2-Amino-4-cyano-phenol

A181547-1G A181547-250MG
ALA181547-1GEA 149.06 USD
ALA181547-1G ALA181547250MG
3-Amino-4-hydroxybenzonitrile ≥98%
3-Amino-4-hydroxybenzonitrile
Formula: C₇H₆N₂O
MW: 134.14 g/mol
Melting Pt: 151-153°C
MDL Number: MFCD00234273
CAS Number: 14543-43-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff76ba11be537a9e0cb322db16db762a7b325e0&vid=8ff76ba11be537a9e0cb322db16db762a7b325e0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR