2-Amino-5-bromo-4-fluorobenzoic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Bromo-2-carboxy-5-fluoroaniline, 5-Bromo-4-fluoroanthranilic acid

A181503-100G A181503-1G A181503-25G A181503-5G
ALA181503-100GEA 467.31 USD
ALA181503-100G ALA181503-1G ALA181503-25G ALA181503-5G
2-Amino-5-bromo-4-fluorobenzoic acid ≥97%
2-Amino-5-bromo-4-fluorobenzoic acid
Formula: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD06409112
CAS Number: 143945-65-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb70886e06a99475a6e24ec4c09d50b78849be2c&vid=eb70886e06a99475a6e24ec4c09d50b78849be2c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR