3-Amino-5-hydroxybenzoic acid hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-Amino-5-hydroxybenzoic acid, HCl

A181460-5G A181460-1G A181460-25G
ALA181460-5GEA 65.46 USD
ALA181460-5G ALA181460-1G ALA181460-25G
3-Amino-5-hydroxybenzoic acid hydrochloride ≥95%
3-Amino-5-hydroxybenzoic acid hydrochloride
Formula: C₇H₈ClNO₃
MW: 189.6000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00043420
CAS Number: 14206-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbd91e33cc1c3631dee452d013579633f7ac1921&vid=fbd91e33cc1c3631dee452d013579633f7ac1921&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR