3-Aminopiperidin-2-one hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Amino-2-piperidone hydrochloride, 3-Aminopiperidin-2-one, HCl, 3-Amino-2-piperidinone hydrochloride

A181382-250MG A181382-500MG A181382-5G
ALA181382250MGEA 18.91 USD
ALA181382250MG ALA181382500MG ALA181382-5G
3-Aminopiperidin-2-one hydrochloride ≥95%
3-Aminopiperidin-2-one hydrochloride
Formula: C₅H₁₁ClN₂O
MW: 150.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09028970
CAS Number: 138377-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e44712a036722681b50d0069323398a9e7e3c28&vid=7e44712a036722681b50d0069323398a9e7e3c28&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR