Methyl-4-aminobicyclo[2.2.2]octane-1-carboxylate ≥98%

Supplier: Aladdin Scientific
Warning

A181289-100MG A181289-1G A181289-250MG A181289-5G
ALA181289100MGEA 187.06 USD
ALA181289100MG ALA181289-1G ALA181289250MG ALA181289-5G
Methyl-4-aminobicyclo[2.2.2]octane-1-carboxylate ≥98%
Formula: C₁₀H₁₇NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD20693756
CAS Number: 135908-33-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9335537b155a70ca44cddf9d792b8cb6fc72d471&vid=9335537b155a70ca44cddf9d792b8cb6fc72d471&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR