2-Amino-2-(4-methylphenyl)acetic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A181169-1G A181169-250MG
ALA181169-1GEA 168.06 USD
ALA181169-1G ALA181169250MG
2-Amino-2-(4-methylphenyl)acetic acid ≥98%
2-Amino-2-(4-methylphenyl)acetic acid
Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00665357
CAS Number: 13227-01-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea3856fa401cfdb27be3875e19a396dd113c5503&vid=ea3856fa401cfdb27be3875e19a396dd113c5503&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR