2-Amino-3,5-difluorobenzoic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A180949-1G A180949-250MG A180949-5G
ALA180949-1GEA 19.86 USD
ALA180949-1G ALA180949250MG ALA180949-5G
2-Amino-3,5-difluorobenzoic acid ≥95%
2-Amino-3,5-difluorobenzoic acid
Formula: C₇H₅F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD01569541
CAS Number: 126674-78-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a8de48730b0c6fd09f91d58701bf57e4be6e3b5&vid=4a8de48730b0c6fd09f91d58701bf57e4be6e3b5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR