3-Amino-2-bromo-6-methylpyridine ≥98%

Supplier: Aladdin Scientific
Warning

A180939-1G A180939-25G A180939-5G
ALA180939-1GEA 55.96 USD
ALA180939-1G ALA180939-25G ALA180939-5G
3-Amino-2-bromo-6-methylpyridine ≥98%
3-Amino-2-bromo-6-methylpyridine
Formula: C₆H₇BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD03095078
CAS Number: 126325-53-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67d3124d72a0df4c176fbc8d7cf03175f4a61a5d&vid=67d3124d72a0df4c176fbc8d7cf03175f4a61a5d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR