3-Amino-2-bromo-4-methylpyridine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-Amino-2-bromo-4-picoline

A180938-100G A180938-1G A180938-25G A180938-5G
ALA180938-100GEA 454.01 USD
ALA180938-100G ALA180938-1G ALA180938-25G ALA180938-5G
3-Amino-2-bromo-4-methylpyridine ≥97%
3-Amino-2-bromo-4-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD04112497
CAS Number: 126325-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a20b60509cec5d0016a6874f535ebca79c05a339&vid=a20b60509cec5d0016a6874f535ebca79c05a339&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR