7-Aminoquinoline hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A180566-1G A180566-250MG A180566-5G
ALA180566-1GEA 65.46 USD
ALA180566-1G ALA180566250MG ALA180566-5G
7-Aminoquinoline hydrochloride ≥95%
7-Aminoquinoline hydrochloride
Formula: C₉H₈N₂·ClH
Storage Temperature: Refrigerator
CAS Number: 1246556-01-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c351b191e36e4f0121bf916541a794d776a3524&vid=7c351b191e36e4f0121bf916541a794d776a3524&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR