2-Amino-5-bromo-3-ethynylpyrazine ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A180121-1G A180121-250MG A180121-50MG A180121-5G
ALA180121-1GEA 112.01 USD
ALA180121-1G ALA180121250MG ALA180121-50MG ALA180121-5G
2-Amino-5-bromo-3-ethynylpyrazine ≥95%
2-Amino-5-bromo-3-ethynylpyrazine
Formula: C₆H₄BrN₃
Storage Temperature: Ambient
MDL Number: MFCD09999142
CAS Number: 1209289-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad33601aa6aafd8a5266b7b18fa7aa4635c0a7a7&vid=ad33601aa6aafd8a5266b7b18fa7aa4635c0a7a7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR