5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 5-Amino-3-cyano-1-(2,6-dichloro-4-trifluoromethylphenyl)pyrazole, 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-1H-pyrazole-3-carbonitrile, 5-Amino-1-(2,6-dichloro-4-trifluoromethylphenyl)-3-cycano pyrazole

A180074-100G A180074-25G A180074-5G
ALA180074-100GEA 185.16 USD
ALA180074-100G ALA180074-25G ALA180074-5G
5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile ≥98%
5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile
Formula: C₁₁H₅Cl₂F₃N₄
MW: 321.0900 g/mol
Melting Pt: 152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00227537
CAS Number: 120068-79-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7e922630516bda27ff9946c840617aca8e5dfdb&vid=a7e922630516bda27ff9946c840617aca8e5dfdb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR