3-Amino-3-(3-fluorophenyl)propionic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Amino-3-(3-fluorophenyl)propanoic acid

A179831-1G A179831-5G
ALA179831-1GEA 9.41 USD
ALA179831-1G ALA179831-5G
3-Amino-3-(3-fluorophenyl)propionic acid ≥97%
3-Amino-3-(3-fluorophenyl)propionic acid
Formula: C₉H₁₀FNO₂
MW: 183.1796 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00764457
CAS Number: 117391-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b503c08940d4f4bc9d83fbb6a989ecea79ff1d4f&vid=b503c08940d4f4bc9d83fbb6a989ecea79ff1d4f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR