3-Amino-5-carboxybenzeneboronic acid ≥98% (contains varying amounts of Anhydride)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
3-Amino-5-boronobenzoic acid, 3-Amino-5-carboxylphenylboronic acid, 3-Borono-5-carboxyaniline
A179771-1G
A179771-250MG
A179771-5G
ALA179771-1GEA
136.71
USD
ALA179771-1G
ALA179771250MG
ALA179771-5G
3-Amino-5-carboxybenzeneboronic acid ≥98% (contains varying amounts of Anhydride)
3-Amino-5-carboxybenzeneboronic acid
Formula:
C₇H₈BNO₄ Melting Pt: 210-212°C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179466 CAS Number: 116378-40-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...