2-Amino-3-fluorobenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A179724-100G A179724-1G A179724-25G A179724-5G
ALA179724-100GEA 10362.51 USD
ALA179724-100G ALA179724-1G ALA179724-25G ALA179724-5G
2-Amino-3-fluorobenzonitrile ≥98%
2-Amino-3-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
MDL Number: MFCD07368774
CAS Number: 115661-37-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b00e43bfd6a90aadc82a4194af23c612d8365568&vid=b00e43bfd6a90aadc82a4194af23c612d8365568&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR