2-(4-Aminophenyl)-2-methylpropanenitrile ≥96%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A179708-1G
A179708-250MG
A179708-25G
A179708-5G
ALA179708-1GEA
59.76
USD
ALA179708-1G
ALA179708250MG
ALA179708-25G
ALA179708-5G
2-(4-Aminophenyl)-2-methylpropanenitrile ≥96%
2-(4-Aminophenyl)-2-methylpropanenitrile
Formula:
C₁₀H₁₂N₂ MW: 160.2200 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09909912 CAS Number: 115279-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...