Atorvastatin calcium salt trihydrate ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Calcium (3R,5R)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate trihydrate, Atorvastatin Calcium Salt Trihydrate, Atorvastatin hemicalcium salt sesquihydrate, Atorvastatin calcium

A179410-1G A179410-25G A179410-5G
ALA179410-1GEA 228.86 USD
ALA179410-1G ALA179410-25G ALA179410-5G
Atorvastatin calcium salt trihydrate ≥97%
Atorvastatin calcium salt trihydrate

Atorvastatin Calcium, off white crystalline powder. It is a synthetic selective inhibitor of HMG-CoA reductase. It is insoluble in aqueous solution with pH below 4 and is freely soluble in methanol.It was used as standard in the method development and validation for simultaneous determination of Atorvastatin calcium and Ezetimibe in tablets using UV spectrophotometric, HPLC and HPTLC methods.

Formula: C₆₆H₇₄CaF₂N₄O₁₃
MW: 1209.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09752074
CAS Number: 344423-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=661fc308a6ed12f5d1b957d353c51026a2c730dc&vid=661fc308a6ed12f5d1b957d353c51026a2c730dc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR