Ac-DL-Pro-OH ≥98%
Supplier: ALADDIN SCIENTIFIC
Warning
|
Synonyms:
1-Acetylpyrrolidine-2-carboxylic acid, N-Acetyl-DL-Proline
A179290-100G
A179290-25G
A179290-5G
ALA179290-100GEA
228.86
USD
ALA179290-100G
ALA179290-25G
ALA179290-5G
Ac-DL-Pro-OH ≥98%
Ac-DL-Pro-OH
Formula:
C₇H₁₁NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00063230 CAS Number: 1074-79-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...