3-Acetyl-1H-pyrrole ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A179139-10G A179139-250MG A179139-5G
ALA179139-10GEA 468.26 USD
ALA179139-10G ALA179139250MG ALA179139-5G
3-Acetyl-1H-pyrrole ≥98%
3-Acetyl-1H-pyrrole
Formula: C₆H₇NO
MW: 109.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00067759
CAS Number: 1072-82-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c89d119081d8ac2f4cf6de889dabbb5a06795a8&vid=0c89d119081d8ac2f4cf6de889dabbb5a06795a8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR