Aminomalonic acid ≥95%

Supplier: ALADDIN SCIENTIFIC

A179126-10G A179126-1G A179126-250MG A179126-50MG A179126-5G
ALA179126-10GEA 12341.36 USD
ALA179126-10G ALA179126-1G ALA179126250MG ALA179126-50MG ALA179126-5G
Aminomalonic acid ≥95%
Aminomalonic acid
Formula: C₃H₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00673796
CAS Number: 1068-84-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af6a9f6bbcfc7bf4392872f9e61fd77d5d3f53c4&vid=af6a9f6bbcfc7bf4392872f9e61fd77d5d3f53c4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR