1-Methyl-1H-indol-5-ylamine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 5-Amino-1-n-methylindole, 1-Methyl-1H-indol-5-amine, 5-Amino-1-methyl-1H-indole

A178842-1G A178842-250MG A178842-25G A178842-5G
ALA178842-1GEA 16.06 USD
ALA178842-1G ALA178842250MG ALA178842-25G ALA178842-5G
1-Methyl-1H-indol-5-ylamine ≥98%
1-Methyl-1H-indol-5-ylamine
Formula: C₉H₁₀N₂
MW: 146.19 g/mol
MDL Number: MFCD03839873
CAS Number: 102308-97-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d860557a991051f52a1925681789d32e5d5a4c0&vid=8d860557a991051f52a1925681789d32e5d5a4c0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR