3-Amino-2-phenylpyridine ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A178752-10G
A178752-1G
A178752-5G
ALA178752-10GEA
538.56
USD
ALA178752-10G
ALA178752-1G
ALA178752-5G
3-Amino-2-phenylpyridine ≥97%
3-Amino-2-phenylpyridine
Formula:
C₁₁H₁₀N₂ Storage Temperature: Ambient |
MDL Number:
MFCD04114113 CAS Number: 101601-80-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...