3-Ethynylpyrazin-2-amine ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Ethynyl-2-pyrazinamine, 2-Amino-3-ethynylpyrazine

A178706-1G A178706-250MG A178706-5G
ALA178706-1GEA 215.56 USD
ALA178706-1G ALA178706250MG ALA178706-5G
3-Ethynylpyrazin-2-amine ≥95%
3-Ethynylpyrazin-2-amine

Product Application:2-Amino-3-ethynylpyrazine is used as reactant in the synthetic preparation of 5H-pyrrolo[2,3-b]pyrazine derivatives that are useful as intermediates for preparation of protein kinase inhibitors.

Formula: C6H5N3
Storage Temperature: Ambient
MDL Number: MFCD09999143
CAS Number: 1005349-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d2ae66c050624a570724a48717f7de63b174a9e&vid=1d2ae66c050624a570724a48717f7de63b174a9e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR