2-Amino-5-bromo-4-methyl-3-nitropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A178693-100G A178693-10G A178693-1G A178693-25G
ALA178693-100GEA 2737.81 USD
ALA178693-100G ALA178693-10G ALA178693-1G ALA178693-25G
2-Amino-5-bromo-4-methyl-3-nitropyridine ≥98%
2-Amino-5-bromo-4-methyl-3-nitropyridine
Formula: C₆H₆BrN₃O₂
Melting Pt: 168-172°C
Storage Temperature: Ambient
MDL Number: MFCD00092197
CAS Number: 100367-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d34bf1e496afe45cc31a2b0ce963b030249dc82&vid=9d34bf1e496afe45cc31a2b0ce963b030249dc82&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR