3-Amino-4-bromophenol ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A178691-100G A178691-1G A178691-250MG
ALA178691-100GEA 748.51 USD
ALA178691-100G ALA178691-1G ALA178691250MG
3-Amino-4-bromophenol ≥98%
3-Amino-4-bromophenol
Formula: C₆H₆BrNO
MW: 188.0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09260886
CAS Number: 100367-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be55e498873a25c84305b57b90d02fa7536fbace&vid=be55e498873a25c84305b57b90d02fa7536fbace&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR