5-Amino-2-(methylthio)pyrimidine-4-carboxylic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION

A178673-1G A178673-250MG
ALA178673-1GEA 75.91 USD
ALA178673-1G ALA178673250MG
5-Amino-2-(methylthio)pyrimidine-4-carboxylic acid ≥97%
5-Amino-2-(methylthio)pyrimidine-4-carboxylic acid
Formula: C₆H₇N₃O₂S
MW: 185.2000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12407810
CAS Number: 100130-05-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52cf8905ffd1597bb84121c2036e638c69360a6f&vid=52cf8905ffd1597bb84121c2036e638c69360a6f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR