2-Amino-5-bromoisonicotinic acid ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A178641-1G
A178641-5G
ALA178641-1GEA
23.66
USD
ALA178641-1G
ALA178641-5G
2-Amino-5-bromoisonicotinic acid ≥98%
2-Amino-5-bromoisonicotinic acid
Formula:
C₆H₅BrN₂O₂ MW: 217.0200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09878357 CAS Number: 1000339-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...