2-Amino-5-pyrimidineboronic acid ≥97% (contains varying amounts of Anhydride)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Aminopyrimidine-5-boronic acid, (2-Aminopyrimidin-5-yl)boronic acid

A178323-1G A178323-25G A178323-5G
ALA178323-1GEA 52.16 USD
ALA178323-1G ALA178323-25G ALA178323-5G
2-Amino-5-pyrimidineboronic acid ≥97% (contains varying amounts of Anhydride)
2-Amino-5-pyrimidineboronic acid

Product Application:2-Aminopyrimidine-5-boronic acid is used as pharmaceutical intermediates.

Formula: C₄H₆BN₃O₂
MW: 138.92 g/mol
Melting Pt: 211 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07375147
CAS Number: 936250-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d824f0ffba914584f7d0a66f2a66ac80e94225d&vid=8d824f0ffba914584f7d0a66f2a66ac80e94225d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR