Azepan-3-ylmethanol hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION

A177665-100MG A177665-500MG
ALA177665100MGEA 508.16 USD
ALA177665100MG ALA177665500MG
Azepan-3-ylmethanol hydrochloride ≥97%
Azepan-3-ylmethanol hydrochloride
Formula: C₇H₁₆ClNO
MW: 165.66 g/mol
Storage Temperature: Refrigerator
CAS Number: 856627-55-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5570e57f6e8a90d33f5379ea27adc7d237d6e8be&vid=5570e57f6e8a90d33f5379ea27adc7d237d6e8be&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR