Alogliptin benzoate ≥99%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
SYR-322, Nesina, (R)-2-((6-(3-Aminopiperidin-1-yl)-3-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)benzonitrile benzoate, SYR322, 2-[[6-[(3R)-3-Amino-1-piperidinyl]-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl]methyl]benzonitrile benzoate, SYR 322, Alogliptin, benzoate, Alogliptin(SYR-322)benzoate, Alogliptin Benzoate, 2-[[6-[(3R)-3-aminopiperidin-1-yl]-3-methyl-2,4-dioxopyrimidin-1-yl]methyl]benzonitrile benzoic acid
Alogliptin is a potent, selective inhibitor of DPP-4 with IC50 of <10 nM, exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9
Formula:
C₁₈H₂₁N₅O₂·C₇H₆O₂ Storage Temperature: Freezer |
MDL Number:
MFCD09833195 CAS Number: 850649-62-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...