2-Amino-6-methylnicotinic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-amino-6-methylpyridine-3-carboxylic acid, 2-Amino-6-methylnicotinic acid

A177594-10G A177594-1G A177594-250MG A177594-5G
ALA177594-10GEA 649.71 USD
ALA177594-10G ALA177594-1G ALA177594250MG ALA177594-5G
2-Amino-6-methylnicotinic acid ≥97%
2-Amino-6-methylnicotinic acid
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 339.9°C
Flash Pt: 159.4°C
Storage Temperature: Ambient
MDL Number: MFCD03427668
CAS Number: 846021-26-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c49a7f2c220a825770f5a8a72c8ce4f1ffeec3e4&vid=c49a7f2c220a825770f5a8a72c8ce4f1ffeec3e4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR