1-Azaspiro[4.5]decane-2,8-dione ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A177361-100MG A177361-1G A177361-250MG A177361-25MG A177361-5G
ALA177361100MGEA 52.16 USD
ALA177361100MG ALA177361-1G ALA177361250MG ALA177361-25MG ALA177361-5G
1-Azaspiro[4.5]decane-2,8-dione ≥97%
1-Azaspiro[4.5]decane-2,8-dione
Formula: C₉H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD14706827
CAS Number: 749861-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32c97fb2ff404040e39e3a4a1d68296211a3659c&vid=32c97fb2ff404040e39e3a4a1d68296211a3659c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR