4-Amino-3,5-dimethylbenzonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A177356-1G A177356-250MG A177356-25G A177356-50MG A177356-5G
ALA177356-1GEA 18.91 USD
ALA177356-1G ALA177356250MG ALA177356-25G ALA177356-50MG ALA177356-5G
4-Amino-3,5-dimethylbenzonitrile ≥97%
4-Amino-3,5-dimethylbenzonitrile
Formula: C₉H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD07369841
CAS Number: 74896-24-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcf37452265b85e683cdfd401f3bcbfbf76654a0&vid=fcf37452265b85e683cdfd401f3bcbfbf76654a0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR