6-Aminonicotinaldehyde 97
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
6-aminopyridine-3-carbaldehyde, 6-Aminonicotinaldehyde
A177224-100MG
A177224-1G
A177224-250MG
A177224-5G
ALA177224100MGEA
10.36
USD
ALA177224100MG
ALA177224-1G
ALA177224250MG
ALA177224-5G
6-Aminonicotinaldehyde 97
6-Aminonicotinaldehyde
Formula:
C₆H₆N₂O MW: 122.125 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09757481 CAS Number: 69879-22-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...