4-(Aminomethyl)tetrahydro-2H-pyran-4-ol hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-(aminomethyl)oxan-4-ol hydrochloride

A177118-100MG A177118-1G A177118-250MG A177118-5G
ALA177118100MGEA 17.96 USD
ALA177118100MG ALA177118-1G ALA177118250MG ALA177118-5G
4-(Aminomethyl)tetrahydro-2H-pyran-4-ol hydrochloride ≥97%
4-(Aminomethyl)tetrahydro-2H-pyran-4-ol hydrochloride
Formula: C₆H₁₄ClNO₂
MW: 167.6339 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18072019
CAS Number: 666261-01-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a353957897256570e966a9a61c23f8b2087d044a&vid=a353957897256570e966a9a61c23f8b2087d044a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR