3-O-Benzyl-4-(hydroxymethyl-1,2-O-isopropylidene)-α-D-erythropentofuranose ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 3-O-Benzyl-4-(hydroxymethyl-1,2-O-isopropylidene)-alpha-D-erythropentofuranose, [(3aR,6S,6aR)-6-(benzyloxy)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-5-yl]methanol, 3-O-Benzyl-4-(hydroxymethyl)-1,2-O-isopropylidene-ALPHA-D-ribofuranose, ((3aR,6S,6aR)-6-(Benzyloxy)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxole-5,5-diyl)dimethanol

A177011-1G A177011-250MG A177011-25G A177011-5G
ALA177011-1GEA 25.56 USD
ALA177011-1G ALA177011250MG ALA177011-25G ALA177011-5G
3-O-Benzyl-4-(hydroxymethyl-1,2-O-isopropylidene)-α-D-erythropentofuranose ≥97%
3-O-Benzyl-4-(hydroxymethyl-1,2-O-isopropylidene)-α-D-erythropentofuranose
Formula: C₁₆H₂₂O₆
MW: 310.342 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08704144
CAS Number: 63593-03-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c52569459700f199e9c8c0a814a65312093f847d&vid=c52569459700f199e9c8c0a814a65312093f847d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR