4-Amino-2-hydroxypyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-Amino-2-pyridone, 4-Aminopyridin-2(1H)-one, 4-Amino-2(1H)-pyridinone, 4-Amino-2-pyridinol, 4-Amino-pyridin-2-ol, 4-amino-1,2-dihydropyridin-2-one, 3-Deazacytosine, 4-Amino-1H-pyridin-2-one

A176346-1G A176346-250MG A176346-25G A176346-5G
ALA176346-1GEA 26.51 USD
ALA176346-1G ALA176346250MG ALA176346-25G ALA176346-5G
4-Amino-2-hydroxypyridine ≥97%
4-Amino-2-hydroxypyridine

Product Application:4-Amino-2-pyridone is used in the preparation of nucleoside analogues with antitumor and antiviral activity.

Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD02682913
CAS Number: 38767-72-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8f30a6cdb3267b5f30ef288693f0708beb0ce2c&vid=b8f30a6cdb3267b5f30ef288693f0708beb0ce2c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR