2-Amino-4-cyanopyrimidine ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning
|
Synonyms:
2-Aminopyrimidine-4-carbonitrile
A176265-100MG
A176265-1G
A176265-250MG
ALA176265100MGEA
31.26
USD
ALA176265100MG
ALA176265-1G
ALA176265250MG
2-Amino-4-cyanopyrimidine ≥97%
2-Amino-4-cyanopyrimidine
Formula:
C₅H₄N₄ MW: 120.1100 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09834971 CAS Number: 36314-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...