3-Aminopyrazine-2-carbaldehyde ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-aminopyrazine-2-carbaldehyde

A176139-50MG A176139-100MG A176139-1G A176139-250MG
ALA176139-50MGEA 51.21 USD
ALA176139-50MG ALA176139100MG ALA176139-1G ALA176139250MG
3-Aminopyrazine-2-carbaldehyde ≥97%
3-Aminopyrazine-2-carbaldehyde
Formula: C5H5N3O
MW: 123.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10696886
CAS Number: 32710-14-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=913472b7bf908ee98f0e67160c7f9b9bffafafa4&vid=913472b7bf908ee98f0e67160c7f9b9bffafafa4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR