7-Azabicyclo[2.2.1]heptane hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A175977-100MG A175977-10G A175977-1G A175977-250MG A175977-50MG A175977-5G
ALA175977100MGEA 27.46 USD
ALA175977100MG ALA175977-10G ALA175977-1G ALA175977250MG ALA175977-50MG ALA175977-5G
7-Azabicyclo[2.2.1]heptane hydrochloride ≥97%
7-Azabicyclo[2.2.1]heptane hydrochloride
Formula: C₆H₁₂ClN
Storage Temperature: Ambient
MDL Number: MFCD00154183
CAS Number: 27514-07-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fddbfd246829f1b6f35b53c249895287930b17a1&vid=fddbfd246829f1b6f35b53c249895287930b17a1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR