Azetidine-2-carboxylic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: azetidine-2-carboxylic acid

A175896-1G A175896-250MG
ALA175896-1GEA 389.41 USD
ALA175896-1G ALA175896250MG
Azetidine-2-carboxylic acid ≥97%
Azetidine-2-carboxylic acid
Formula: C₄H₇NO₂
MW: 101.11 g/mol
MDL Number: MFCD00066660
CAS Number: 2517-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR