3-Aminopiperidine-2,6-dione hydrochloride ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
3-Aminopiperidine-2,6-dione hydrochloride is a reagent for preparing lenalidomide that can induce ubiquitination and degradation of CK1α in del(5q) MDS. It can also be used to prepare phthalimide conjugates that can promote ligand-dependent target protein degradation.
Formula:
C5H9ClN2O2 Storage Temperature: Refrigerator |
MDL Number:
MFCD11042437 CAS Number: 24666-56-6 EINECS: 806-697-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...