3-Amino-1-methyl-1H-pyrazole-4-carbonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A175719-100MG A175719-1G A175719-250MG
ALA175719100MGEA 102.51 USD
ALA175719100MG ALA175719-1G ALA175719250MG
3-Amino-1-methyl-1H-pyrazole-4-carbonitrile ≥97%
3-Amino-1-methyl-1H-pyrazole-4-carbonitrile
Formula: C5H6N4
Storage Temperature: Ambient
CAS Number: 21230-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea8255feb8bf74d72c501ea35c1db4e24aaa6481&vid=ea8255feb8bf74d72c501ea35c1db4e24aaa6481&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR