1-(Aminomethyl)cyclobutanol ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 1-(aminomethyl)cyclobutan-1-ol

A175135-1G A175135-250MG A175135-50MG A175135-5G
ALA175135-1GEA 120.56 USD
ALA175135-1G ALA175135250MG ALA175135-50MG ALA175135-5G
1-(Aminomethyl)cyclobutanol ≥97%
1-(Aminomethyl)cyclobutanol
Formula: C₅H₁₁NO
MW: 101.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11040537
CAS Number: 180205-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fc4ce598f82f1c16e7afda69ad516cdfa6bfa82&vid=7fc4ce598f82f1c16e7afda69ad516cdfa6bfa82&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR