2-Amino-5,6-dihydro-4H-benzothiazol-7-one ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-5,6-dihydrobenzo[d]thiazol-7(4H)-one

A175001-1G A175001-250MG A175001-25G A175001-5G
ALA175001-1GEA 15.11 USD
ALA175001-1G ALA175001250MG ALA175001-25G ALA175001-5G
2-Amino-5,6-dihydro-4H-benzothiazol-7-one ≥97%
2-Amino-5,6-dihydro-4H-benzothiazol-7-one
Formula: C₇H₈N₂OS
MW: 168.22 g/mol
MDL Number: MFCD00460519
CAS Number: 17583-10-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd600caa23049ee0593c31b169a72e93e816580c&vid=cd600caa23049ee0593c31b169a72e93e816580c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR