3-Azetidinone hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Azetidin-3-one hydrochloride

A174998-1G A174998-250MG
ALA174998-1GEA 16.06 USD
ALA174998-1G ALA174998250MG
3-Azetidinone hydrochloride ≥97%
3-Azetidinone hydrochloride
Formula: C₃H₆ClNO
MW: 107.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861742
CAS Number: 17557-84-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=293e91d584f470f4fa8dce6ce8920d643c8d8e72&vid=293e91d584f470f4fa8dce6ce8920d643c8d8e72&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR