AZD-8186 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC

A174656-50MG A174656-5MG A174656-25MG
ALA174656-50MGEA 402.71 USD
ALA174656-50MG ALA174656-5MG ALA174656-25MG
AZD-8186 ≥98%, Moligand™
AZD-8186
Formula: C₂₄H₂₅F₂N₃O₄
Storage Temperature: Freezer
MDL Number: MFCD27987908
CAS Number: 1627494-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cbf98ac04cd6bb6fe7807ebffb9708d8159be9f&vid=2cbf98ac04cd6bb6fe7807ebffb9708d8159be9f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR