6-Aminotetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-ol ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: Ticagrelor Related Compound 1, (3aR,4S,6R,6aS)-6-Amino-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-ol, (3aR,4S,6R,6aS)-6-amino-2,2-dimethyl-hexahydrocyclopenta[d][1,3]dioxol-4-ol

A174541-100G A174541-100MG A174541-10G A174541-1G A174541-250MG A174541-25G A174541-5G
ALA174541-100GEA 429.31 USD
ALA174541-100G ALA174541100MG ALA174541-10G ALA174541-1G ALA174541250MG ALA174541-25G ALA174541-5G
6-Aminotetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-ol ≥97%
6-Aminotetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-ol
Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20527303
CAS Number: 155899-66-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=816dc806c364869d7afae8672534c32b56235b22&vid=816dc806c364869d7afae8672534c32b56235b22&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR