2-(Azetidin-3-yl)ethan-1-ol hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A174496-1G A174496-250MG
ALA174496-1GEA 4437.36 USD
ALA174496-1G ALA174496250MG
2-(Azetidin-3-yl)ethan-1-ol hydrochloride ≥97%
2-(Azetidin-3-yl)ethan-1-ol hydrochloride
Formula: C₅H₁₂ClNO
MW: 137.61 g/mol
Storage Temperature: Ambient
CAS Number: 152537-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c994c9bb34278fb94e353acf12d64c21bbb14fd&vid=0c994c9bb34278fb94e353acf12d64c21bbb14fd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR