2-(3-Aminooxetan-3-yl)ethan-1-ol hemioxalate ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION

A174483-100MG A174483-1G A174483-250MG A174483-500MG
ALA174483100MGEA 323.86 USD
ALA174483100MG ALA174483-1G ALA174483250MG ALA174483500MG
2-(3-Aminooxetan-3-yl)ethan-1-ol hemioxalate ≥97%
2-(3-Aminooxetan-3-yl)ethan-1-ol hemioxalate
Formula: C₁₂H₂₄N₂O₈
MW: 324.33 g/mol
Storage Temperature: Ambient
CAS Number: 1523618-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=099536fe3a759d15e4fde30888311206aad68bb3&vid=099536fe3a759d15e4fde30888311206aad68bb3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR